Learn More
Selleck Chemical LLC Vinorelbine Tartrate 50mg 125317-39-7
SDP
Supplier:  Selleck Chemical LLC S426950MG
Encompass_Preferred
Vinorelbine Tartrate is a semi-synthetic vinca alkaloid, and inhibits mitosis through interaction with tubulin. Vinorelbine Tartrate exhibits anti-tumor activities via inducing the mitotic apoptosis, autophagy and inflammation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3825