Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinorelbine tartrate, 98%

Catalog No. AC461980250
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461980250 25 mg
AC461981000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461980250 Supplier Thermo Scientific Chemicals Supplier No. 461980250
Add to Cart

CAS: 125317-39-7 | C53H66N4O20 | 1079.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125317-39-7
Molecular Formula C53H66N4O20
Molecular Weight (g/mol) 1079.12
InChI Key CILBMBUYJCWATM-NPJYPKOYSA-N
IUPAC Name bis((2R,3R)-2,3-dihydroxybutanedioic acid); methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(12S)-16-ethyl-12-(methoxycarbonyl)-1,10-diazatetracyclo[12.3.1.0³,¹¹.0⁴,⁹]octadeca-3(11),4,6,8,15-pentaen-12-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate
SMILES O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CCC1=CC2CN(C1)CC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC

Specifications

Quantity 25 mg
Formula Weight 1079.12
Chemical Name or Material Vinorelbine tartrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Causes skin irritation.

Causes serious eye irritation.

Suspected of causing genetic defects.

May damage fertility.

May damage the unborn child.

Causes damage to organs through
Precautionary Statement GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

IF ON SKIN: Wash with plenty of soap and water.

If skin irritation occurs: Get medical advice/attention.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.