Learn More
Cayman Chemical VInorelbIntartrate 50mg
SDP
Catalog No. 502981481
Encompass_Preferred
An inhibitor of microtubule polymerization that blocks mitosis at the metaphase/anaphase transition, preventing HeLa cell proliferation (IC50 = 1.25 nM)
Catalog No. 50-298-1481 Supplier Cayman Chemical Supplier No. 2126250 MG
An inhibitor of microtubule polymerization that blocks mitosis at the metaphase/anaphase transition, preventing HeLa cell proliferation (IC50 = 1.25 nM)