Learn More
Cayman Chemical VInpocetIn 100mg
SDP
Catalog No. 502985471
Encompass_Preferred
An inhibitor of PDE1 that has diverse cerebral and neurological effects in vivo; also directly inhibits IKKβ (IC50 = 17 µM in a cell-free system) and blocks TNF-α- and LPS-mediated activation of NF-κB in cells and in vivo
Catalog No. 50-298-5471 Supplier Cayman Chemical Supplier No. 19578100 MG
An inhibitor of PDE1 that has diverse cerebral and neurological effects in vivo; also directly inhibits IKKβ (IC50 = 17 µM in a cell-free system) and blocks TNF-α- and LPS-mediated activation of NF-κB in cells and in vivo