Learn More
Enzo Life Sciences Vinpocetine (100mg). CAS: 42971-09-5
SDP
Supplier:  Enzo Life Sciences BMLPD1850100
Encompass_Preferred
Selective inhibitor of Ca2+/calmodulin-dependent phosphodiesterase (PDE I, IC50=20 µM). Purity: ≥98% (Assay). Solubility: Soluble in DMSO or acetone. Long Term Storage: Ambient.
Catalog No. 50-201-1393