Learn More
Apexbio Technology LLC Vinpocetine 42971-09-5 100mg
SDP
Catalog No. 502240828
Encompass_Preferred
Vinpocetine (CAS 42971-09-5) is a selective inhibitor of cyclic GMP phosphodiesterase (PDE) exhibiting a Ki of 14 2 M for Ca2 -dependent PDE in vitro It acts as a non-competitive inhibitor effectively modulating intracellular cGMP levels in smooth muscle In rat aortic cytosolic preparations vinpocetine inhibits Ca2 -PDE activity significantly while demonstrating minimal inhibition of Ca2 -independent cGMP PDE and cAMP PDE (Ki values 1000 M) Additionally vinpocetine attenuates 5-HT-induced contractions in intact rat aorta with an EC50 of 1 M This compound is used in research to explore mechanisms of smooth muscle relaxation and PDE regulation
This item is not returnable. View return policy
Catalog No. 50-224-0828 Supplier Apexbio Technology LLC Supplier No. B2276100
Vinpocetine (CAS 42971-09-5) is a selective inhibitor of cyclic GMP phosphodiesterase (PDE) exhibiting a Ki of 14 2 M for Ca2 -dependent PDE in vitro It acts as a non-competitive inhibitor effectively modulating intracellular cGMP levels in smooth muscle In rat aortic cytosolic preparations vinpocetine inhibits Ca2 -PDE activity significantly while demonstrating minimal inhibition of Ca2 -independent cGMP PDE and cAMP PDE (Ki values 1000 M) Additionally vinpocetine attenuates 5-HT-induced contractions in intact rat aorta with an EC50 of 1 M This compound is used in research to explore mechanisms of smooth muscle relaxation and PDE regulation