Learn More
AdipoGen Vinpocetine (20 mg)
SDP
Supplier:  AdipoGen AGCN20454M020
Encompass_Preferred
Chemical. CAS: 42971-09-5. Formula: C22H26N2O2. MW: 350.5. Semi-synthetic from Voacanga africana. Selective Ca2+-calmodulin dependent cGMP-phosphodiesterase (PDE1) inhibitor. Shows vasorelaxant activity. Neuroprotective agent. Selectively inhibits voltage-sensitive 2+ channels Potent anti-inflammatory agent. Inhibitor of NF-kappaB-dependent inflammatory responses by directly targeting IKK. Shown to inhibit the NLRP3 inflammasome. Antioxidant. Free radical scavenger. Anticancer compound. Anticonvulsant.
Catalog No. 50-205-3621