Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl 10-Undecenoate (stabilized with MEHQ) 92.0+%, TCI America™
SDP

Catalog No. U005725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U005725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U005725ML Supplier TCI America Supplier No. U005725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5299-57-0
Molecular Formula C13H22O2
Molecular Weight (g/mol) 210.32
MDL Number MFCD00142351
InChI Key HUGGPHJJSYXCDJ-UHFFFAOYSA-N
Synonym 10-Undecenoic Acid Vinyl Ester
PubChem CID 544393
IUPAC Name ethenyl undec-10-enoate
SMILES C=CCCCCCCCCC(=O)OC=C

Specifications

Color Yellow
Boiling Point 127°C
Quantity 25 mL
Formula Weight 210.32
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl 10-Undecenoate (stabilized with MEHQ)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.