Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl 4-tert-Butylbenzoate 98.0+%, TCI America™
SDP

Catalog No. B138625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B138625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B138625ML Supplier TCI America Supplier No. B138625ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15484-80-7
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00080692
InChI Key ZLHVSEPPILCZHH-UHFFFAOYSA-N
Synonym 4-tert-Butylbenzoic Acid Vinyl Ester
PubChem CID 84940
IUPAC Name ethenyl 4-tert-butylbenzoate
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)OC=C

Specifications

Color Yellow
Boiling Point 111°C
Quantity 25 mL
Formula Weight 204.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl 4-tert-Butylbenzoate
EINECSNumber (3)-3697
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.