Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl acetate, MilliporeSigma™ Supelco™

Catalog No. 111020125 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0125 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0125 Supplier MilliporeSigma Supelco Supplier No. 48486
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vinyl acetate is an important ester of vinyl alcohol, which can find applications in the production of homo and copolymer dispersions. The production of vinyl acetate involves the addition of acetic acid to acetylene in the gas phase in the presence of zinc acetate/charcoal as catalyst.

Cleaning Products, Cosmetics, Environmental, Food and Beverages, Personal Care and Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 801°F
CAS 108-05-4
Vapor Pressure 88 mmHg (20°C)
Vapor Density 3 (vs. air)
Density 0.934 g/mL (at 25°C)
Flash Point -8°C
Physical Form Neat
Linear Formula CH3 CO2 CH=CH2
MDL Number MFCD00008713
Melting Point -93°C
Boiling Point 72°C to 73°C
Synonym Acetoxyethylene
Type Food and Cosmetic Component Standard
Quantity 1000 mg
Formula Weight 86.09
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.