Learn More
VGuard™ Vinyl Arm Sleeve
Catalog No. 17005965
Click to view available options
Color:
Blue
Catalog No. Color
17-005-965 Blue
1 options
This item is not returnable. View return policy
Catalog No. 17-005-965 Supplier VGuard™ Supplier No. B66O20
This item is not returnable. View return policy
Lightweight and water repellent arm protection.
  • Heavy duty vinyl provides a comfortable fit for all day wear
  • Protection against fats, oils, chemicals, liquids, dirt and grime
  • Resists snags, punctures and abrasion
  • Keeps skin and hair from contaminating product
Specifications
Product Type General Purpose Sleeve
Construction Vinyl
Material Vinyl
Color Blue
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Length (Metric) 457.2 mm