Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vinyl bromide

Catalog No. 503125757 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
2.2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-5757 2.2 kg
50-312-5627 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-312-5757 Supplier Sigma Aldrich Supplier No. V19022.2KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Vinyl bromide

Specifications

CAS 593-60-2
Melting Point -139°C
Density 1.517 g/mL (at 25°C)
Boiling Point 16°C (750 mmHg)
Molecular Formula C2H3Br
Linear Formula CH2=CHBr
Quantity 2.2 kg
MDL Number MFCD00000183
Synonym Bromoethylene
Molecular Weight (g/mol) 106.95
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.