Learn More
Alzet ALZETTM Vinyl Catheter Tubing for Use with any ALZET Osmotic Pump, Pack of 10 Tubes
SDP
Catalog No. NC9166951
Encompass
These vinyl ALZET catheters connect to any ALZET Osmotic Pump to direct delivery of test agents to a local target. EtO sterilized and ready for immediate use.Features•Medical grade•Pre-cut to 15 cm lengths•Individually packaged•Ready to use•Fit all pump models
Catalog No. NC9166951 Supplier Alzet Supplier No. 0007760
These vinyl ALZET catheters connect to any ALZET Osmotic Pump to direct delivery of test agents to a local target. EtO sterilized and ready for immediate use.Features•Medical grade•Pre-cut to 15 cm lengths•Individually packaged•Ready to use•Fit all pump models