Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl Cinnamate (stabilized with MEHQ) 99.0+%, TCI America™
SDP

Catalog No. C0899500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0899500ML 500 mL
C089925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0899500ML Supplier TCI America Supplier No. C0899500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3098-92-8
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.199
MDL Number MFCD00059324
InChI Key WGXGKXTZIQFQFO-CMDGGOBGSA-N
Synonym Cinnamic Acid Vinyl Ester
PubChem CID 5371819
IUPAC Name ethenyl (E)-3-phenylprop-2-enoate
SMILES C=COC(=O)C=CC1=CC=CC=C1

Specifications

Melting Point -28°C
Color Yellow
Boiling Point 125°C
Quantity 500 mL
Formula Weight 174.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Cinnamate (stabilized with MEHQ)
EINECSNumber (3)-4655
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.