Learn More
Electron Microscopy Sciences VIKEM Vinyl Vinyl Coated Lead Wire
SDP
Catalog No. 5029662
Encompass_Preferred
This lead wire is 1/4" (6.4 mm) in diameter and is coated with a chemical-resistant red Vikem vinyl. Its 12" (300 mm) length and its flexibility make it a very versatile weight. It can be coiled around a flask, beaker, or any other irregular shape. It weighs only 1/4 lb (114g).
Catalog No. 50-296-62 Supplier Electron Microscopy Sciences Supplier No. 6444300
This lead wire is 1/4" (6.4 mm) in diameter and is coated with a chemical-resistant red Vikem vinyl. Its 12" (300 mm) length and its flexibility make it a very versatile weight. It can be coiled around a flask, beaker, or any other irregular shape. It weighs only 1/4 lb (114g).