Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl Crotonate (stabilized with MEHQ) 99.0+%, TCI America™
SDP

Catalog No. C0897500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0897500ML 500 mL
C089725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0897500ML Supplier TCI America Supplier No. C0897500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14861-06-4
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00009288
InChI Key IYNRVIKPUTZSOR-HWKANZROSA-N
Synonym vinyl crotonate, vinyl 2-butenoate, crotonic acid, vinyl ester, crotonic acid vinyl ester, unii-smc9a01nvs, ethenyl e-but-2-enoate, vinylester kyseliny krotonove, vinyl but-2-enoate, smc9a01nvs, vinylester kyseliny krotonove czech
PubChem CID 5365800
IUPAC Name ethenyl (E)-but-2-enoate
SMILES CC=CC(=O)OC=C

Specifications

Melting Point -60°C
Color Colorless
Boiling Point 134°C
Quantity 500 mL
UN Number 3272
Formula Weight 112.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Crotonate (stabilized with MEHQ)
EINECSNumber (2)-4046
RTECSNumber GQ5850000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.