Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Vinyl Duct Tape

Catalog No. 19072109 Shop All 3M Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-072-109 Yellow
19-072-110 Gray
19-072-111 Green
19-072-115 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-072-109 Supplier 3M™ Supplier No. 7100146009
Add to Cart
Add to Cart

3M™ Vinyl Duct Tape is a hand-tearable general purpose tape with a rubber adhesive and embossed vinyl backing.

  • Rubber adhesive sticks fast and stays fast on many surfaces, even irregular surfaces
  • Embossed vinyl backing seals out dust, dirt, and moisture, and resists abrasion and many chemicals
  • Ideal for a variety of purposes including temporary repairs, identification, contrast, decoration, securing wiring, bundling, sealing, holding, protecting, and other jobs including applications requiring moisture resistance
  • Peels off cleanly after 30 minutes on painted steel as hot as 200°F (93°C)
  • Easy to tear and conforms to irregular surfaces
  • Available in a variety of colors for coding, process tagging, highlighting trip hazards, and enhancing décor
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Product Type Vinyl Duct Tape
For Use With (Application) Communications, Design and Construction, Manufacturing, Mining, Oil and Gas, Transportation
Model 3903
Quantity 24/Cs.
Thickness (English) 6.5 mil
Dimensions (L x W) 50 yd. x 2 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.