Learn More
MSC VINYL DUCT TAPE
Catalog No. 5000795765
Encompass
This vinyl duct tape features a durable rubber adhesive, designed for reliable performance in various applications.
  • Durable vinyl construction
  • Rubber adhesive for strong hold
  • Provides 6.3 mil thickness for robust use
  • Offers 14 lb/in tensile strength
  • Yellow color for visibility
  • Operates effectively up to 250°F
Catalog No. 50-007-95765 Supplier MSC Supplier No. 20705026
This vinyl duct tape features a durable rubber adhesive, designed for reliable performance in various applications.
  • Durable vinyl construction
  • Rubber adhesive for strong hold
  • Provides 6.3 mil thickness for robust use
  • Offers 14 lb/in tensile strength
  • Yellow color for visibility
  • Operates effectively up to 250°F