Learn More
MSC VINYL DUCT TAPE
Catalog No. 5000688937
Encompass
This duct tape features a rubber adhesive, providing 14 lb/in tensile strength, and comes in black. It is suitable for a wide range of general-purpose applications.
  • Duct tape type
  • Thickness of 6.3 mil
  • Width of 3 in.
  • Black color
  • Length of 50 yd
  • Rubber adhesive material
  • Vinyl tape material
  • Tensile strength of 14 lb/in
  • Maximum operating temperature of 250°F
Catalog No. 50-006-88937 Supplier MSC Supplier No. 20709184
This duct tape features a rubber adhesive, providing 14 lb/in tensile strength, and comes in black. It is suitable for a wide range of general-purpose applications.
  • Duct tape type
  • Thickness of 6.3 mil
  • Width of 3 in.
  • Black color
  • Length of 50 yd
  • Rubber adhesive material
  • Vinyl tape material
  • Tensile strength of 14 lb/in
  • Maximum operating temperature of 250°F