Learn More
Beckman Coulter 96 Well Plate for use with Vi-CELL BLU, 5/pk
Catalog No. NC1695434
Encompass_Preferred
Quantity of 5, 96 well plates for use with Vi-CELL BLU. Use the plate cover slip, C24842, with the plate.
Catalog No. NC1695434 Supplier Beckman Coulter Supplier No. C99716
Quantity of 5, 96 well plates for use with Vi-CELL BLU. Use the plate cover slip, C24842, with the plate.