Learn More
MSC Bertech Anti-Fatigue Mat, 3x3 ft, 3/8 in, 1 ea
Catalog No. 5000688399
Encompass
This anti-fatigue mat is designed with a textured surface for dry areas. It features beveled edges, providing comfort and is suitable for light-duty traffic environments.
  • Beveled edges for safety and comfort
  • Suitable for light-duty traffic
  • Textured surface for enhanced grip
  • Slip-resistant design
  • Ideal for dry environments
  • Constructed from durable vinyl material
Catalog No. 50-006-88399 Supplier MSC Supplier No. 18395681
This anti-fatigue mat is designed with a textured surface for dry areas. It features beveled edges, providing comfort and is suitable for light-duty traffic environments.
  • Beveled edges for safety and comfort
  • Suitable for light-duty traffic
  • Textured surface for enhanced grip
  • Slip-resistant design
  • Ideal for dry environments
  • Constructed from durable vinyl material