Learn More
Electron Microscopy Sciences Disposable Vinyl Gloves, Large, 245 mm length
SDP
Catalog No. 50365636
Encompass_Preferred
Medical grade non-sterile gloves are made from synthetic polyvinyl chloride and are powder-free, seemless, checmical resistant, ambidextrous. These vinyl gloves offer a unique balance of softness flexibility and strength.
Size: Large
Length: 245 mm
This item is not returnable. View return policy
Catalog No. 50-365-636 Supplier Electron Microscopy Sciences Supplier No. 7110830
Medical grade non-sterile gloves are made from synthetic polyvinyl chloride and are powder-free, seemless, checmical resistant, ambidextrous. These vinyl gloves offer a unique balance of softness flexibility and strength.
Size: Large
Length: 245 mm