Learn More
Electron Microscopy Sciences Disposable Vinyl Gloves, Small, 235 mm length
SDP
Supplier:  Electron Microscopy Sciences 7110812
Encompass_Preferred
Medical grade non-sterile gloves are made from synthetic polyvinyl chloride and are powder-free, seemless, checmical resistant, ambidextrous. These vinyl gloves offer a unique balance of softness flexibility and strength.
Size: Small
Length: 235 mm
Catalog No. 50-930-802