Learn More
Enzo Life Sciences Vinyl-L-NIO (25mg). CAS: 728944-69-2
SDP
Catalog No. 502008433
Encompass_Preferred
Very selective and potent neuronal nitric oxide synthase (nNOS/NOS I) inhibitor. Alternative name: N5-(1-Imino-3-butenyl)-L-ornithine, L-VNIO. Formula: C9H17N3O2 . HCl. MW: 235.7. Purity: ≥95% (TLC). Appearance: Off-white solid to clear film. Solubility: Soluble in water, 100% ethanol (30mg/ml), DMSO (50mg/ml) or dimethyl formamide (50mg/ml). Long Term Storage: +4°C. Handling: Very hygroscopic. Open only in anhydrous atmosphere.
Catalog No. 50-200-8433 Supplier Enzo Life Sciences Supplier No. ALX270216M025
Very selective and potent neuronal nitric oxide synthase (nNOS/NOS I) inhibitor. Alternative name: N5-(1-Imino-3-butenyl)-L-ornithine, L-VNIO. Formula: C9H17N3O2 . HCl. MW: 235.7. Purity: ≥95% (TLC). Appearance: Off-white solid to clear film. Solubility: Soluble in water, 100% ethanol (30mg/ml), DMSO (50mg/ml) or dimethyl formamide (50mg/ml). Long Term Storage: +4°C. Handling: Very hygroscopic. Open only in anhydrous atmosphere.