Learn More
Cayman Chemical Vinyl-L-NIO (hydrochloride), Purity: ≥95%, CAS: 728944-69-2, 10 mg
SDP
Catalog No. 502893930
Encompass_Preferred
A potent, selective inhibitor of nNOS with Ki values for inhibition of nNOS, eNOS and iNOS of 100 nM, 12 µM and 60 µM, respectively
Catalog No. 50-289-3930 Supplier Cayman Chemical Supplier No. 8033010 MG
A potent, selective inhibitor of nNOS with Ki values for inhibition of nNOS, eNOS and iNOS of 100 nM, 12 µM and 60 µM, respectively