Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vinyl laurate

Catalog No. 108961KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kg
167 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-896-1KG 1 kg
10-896-167KG 167 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-896-1KG Supplier Sigma Aldrich Supplier No. 108961KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Vinyl laurate is a clear, colorless to light yellow colored liquid
  • The flash point is 125c
  • It forms aqueous colloidal microgel by copolymerization with n-isopropylacrylamide
  • The enzymatic acylation of avicel cellulose with vinyl laurate, a solvent free reaction system, was studied.vinyl laurate was used in the synthesis of laurate and stearate esters of corn starch
  • It was used as the acyl donor during synthesis of sucrose laurate esters catalyzed by bacillus pseudofirmus al-89
  • Vinyl laurate was also used in the preparation of chirally deuterated benzyl chlorides.prices for bulk quantities on request

Specifications

CAS 2146-71-6
Density 0.871 g/mL (at 20°C (literature))
Boiling Point 254°C (1013 hPa (lit.))
Molecular Formula C14H26O2
Linear Formula CH3(CH2)10 COOCH=CH2
MDL Number MFCD00048421
Quantity 1 kg
Synonym VL
Molecular Weight (g/mol) 226.36
Percent Purity ≥98.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.