Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vinyl laurate

Catalog No. 61725250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-725-250ML 250 mL
61-725-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 61-725-250ML Supplier Sigma Aldrich Supplier No. 61725250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Vinyl laurate acts as an acyl donor during the transeterification of L-α-glycerophosphorylcholine (gpc) in the presence of candida antarctica lipase b as catalyst to form lysophosphatidylcholine (lpc).vinyl laurate can be used to synthesize the following:
  • Sucrose laurate esters from sucrose in organic solvents
  • Poly(vinyl laurate) (pvl)latexes
  • Porous conducting soft polymer foams which can be used as gas sensors

Specifications

CAS 2146-71-6
Density 0.871 g/mL (at 20°C (literature))
Boiling Point 254°C (1013 hPa (lit.))
Molecular Formula C14H26O2
Refractive Index n20/D 1.441
Linear Formula CH3(CH2)10 COOCH=CH2
MDL Number MFCD00048421
Quantity 250 mL
Molecular Weight (g/mol) 226.36
Percent Purity ≥99.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.