Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl Laurate, MP Biomedicals

Catalog No. ICN219999 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219999 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219999 Supplier MP Biomedicals, Inc Supplier No. 0521999910

Chemical Identifiers

CAS 2146-71-6
Molecular Formula C14H26O2
Molecular Weight (g/mol) 226.36
InChI Key GLVVKKSPKXTQRB-UHFFFAOYSA-N
Synonym vinyl laurate, dodecanoic acid, ethenyl ester, vinyl dodecanoate, lauric acid, vinyl ester, lauric acid vinyl ester, lauric acid vinyl, acmc-209t6q, dodecanoic acid,ethenyl ester, lauric acid, vinyl ester 8ci, vinyl laurate gc
PubChem CID 75069
IUPAC Name ethenyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC=C

Specifications

Quantity 10 g
Chemical Name or Material Vinyl Laurate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.