Learn More
Safety Zone VINYL MED EX PF SM 1000/CS
Supplier:  Safety Zone GVEPSM1C
Encompass_Preferred
CLEAR VINYL GLOVES MEDICAL GRADE PRODUCED WITH 100% VINYL RESIN POWDER FREE NOT MADE WITH NATURAL RUBBER LATEX 100% DEHP & DINP FREE STANDARD LENGTH (9.25″ +/- 0.25″) SMALL PACKED 100/BX 10BX/CS (1,000 PCS/CS TOTAL)
Catalog No. NC1118584