Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl Palmitate (stabilized with MEHQ) 96.0+%, TCI America™
SDP

Catalog No. P084825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P084825ML 25 mL
P0848500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P084825ML Supplier TCI America Supplier No. P084825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 693-38-9
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.468
MDL Number MFCD00059288
InChI Key UJRIYYLGNDXVTA-UHFFFAOYSA-N
Synonym Palmitic Acid Vinyl Ester
PubChem CID 69658
IUPAC Name ethenyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC=C

Specifications

Color White-Yellow
Quantity 25 mL
Formula Weight 282.47
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Vinyl Palmitate (stabilized with MEHQ)
EINECSNumber (2)-4160
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.