Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl propionate, 97%, stab., Thermo Scientific™

Catalog No. AAL1293130
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1293130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1293130 Supplier Thermo Scientific Chemicals Supplier No. L1293130

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-38-4
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00027004
InChI Key UIWXSTHGICQLQT-UHFFFAOYSA-N
Synonym vinyl propionate, propanoic acid, ethenyl ester, vinyl propanoate, propionic acid vinyl ester, propionic acid, vinyl ester, vinylpropionate, vinylester kyseliny propionove, vinylester kyseliny propionove czech, propanoic acid, ethenyl ester, homopolymer, propionic acid vinyl
PubChem CID 7750
IUPAC Name ethenyl propanoate
SMILES CCC(=O)OC=C

Specifications

Melting Point -80°C
Boiling Point 93°C to 94°C
Flash Point 6°C (42°F)
Linear Formula CH3CH2CO2CH=CH2
Quantity 250 g
UN Number UN3272
Beilstein 1741731
Formula Weight 100.12
Percent Purity 97%
Chemical Name or Material Vinyl propionate, stabilized
Hazard Category Danger
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
RTECSNumber UF8575000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.