Learn More
Grainger Buyers Products Vinyl Tarp, 18 oz, 7-1/2 x 20 ft, 1 ea
Catalog No. 502786449
Encompass_Preferred
This solid, heavy-duty tarp provides secure coverage for dump truck beds. Constructed from 18 oz polyester with a UV-resistant PVC coating, it is designed for demanding environments and is rated for temperatures up to 400°F, making it suitable for covering hot asphalt.
  • Manufactured with 18 oz polyester
  • Features UV-resistant PVC coating
  • Equipped with brass grommets
  • Includes heavy-duty double-stitched and reinforced hems
  • Rated for service temperatures up to 400°F
  • Designed for dump applications
Catalog No. 50-278-6449 Supplier Grainger Supplier No. 19A821
This solid, heavy-duty tarp provides secure coverage for dump truck beds. Constructed from 18 oz polyester with a UV-resistant PVC coating, it is designed for demanding environments and is rated for temperatures up to 400°F, making it suitable for covering hot asphalt.
  • Manufactured with 18 oz polyester
  • Features UV-resistant PVC coating
  • Equipped with brass grommets
  • Includes heavy-duty double-stitched and reinforced hems
  • Rated for service temperatures up to 400°F
  • Designed for dump applications