Learn More
Grainger Buyers Products Solid Tarp, Heavy Duty Vinyl, 7 ft. x 21 ft., 1 ea.
Catalog No. 502785126
Encompass_Preferred
This solid heavy-duty black tarp is designed for secure coverage of dump beds, making it ideal for various dump applications. Constructed from durable 18 oz. polyester with a UV-resistant PVC coating, it is rated for temperatures up to 400°F, allowing for the transport of hot asphalt.
  • Constructed from 18 oz. polyester with a UV-resistant PVC coating
  • Rated to 400°F for hot asphalt applications
  • Features reinforced hems with 2 in. heavy-duty nylon webbing
  • Equipped with brass grommets with a 5/8 in. inner diameter for secure tie-down points
  • Includes tie-down points around the full perimeter
  • Compatible with side-mount and underbody mount tarp systems
  • Heavy-duty double-stitched construction
Catalog No. 50-278-5126 Supplier Grainger Supplier No. 19A818
This solid heavy-duty black tarp is designed for secure coverage of dump beds, making it ideal for various dump applications. Constructed from durable 18 oz. polyester with a UV-resistant PVC coating, it is rated for temperatures up to 400°F, allowing for the transport of hot asphalt.
  • Constructed from 18 oz. polyester with a UV-resistant PVC coating
  • Rated to 400°F for hot asphalt applications
  • Features reinforced hems with 2 in. heavy-duty nylon webbing
  • Equipped with brass grommets with a 5/8 in. inner diameter for secure tie-down points
  • Includes tie-down points around the full perimeter
  • Compatible with side-mount and underbody mount tarp systems
  • Heavy-duty double-stitched construction