Learn More
Brady™ Vinyl Sign Kit: NFPA Placard System Kit - ACID ALK OXY COR
Shop All Brady Corporation ProductsDescription
- Vinyl material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Self sticking vinyl will adhere to difficult low surface energy items, including: PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics
- Excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes.
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Specifications
| Material | Vinyl |
| Mounting Type | Self-adhesive |
| Shape | Diamond |
| Languages | English |
| Dimensions (L x W) | 15 x 15 in. |
| Displayed Text | Vinyl Sign Kit: NFPA Placard System Kit - ACID ALK OXY COR |
| Product Type | Safety Sign |
| Color | White |
Safety and Handling
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.