Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl Stearate (stabilized with MEHQ) 95.0+%, TCI America™
SDP

Catalog No. S0241500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0241500G 500 g
S024125G 25 g
S0241100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S0241500G Supplier TCI America Supplier No. S0241500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111-63-7
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.522
MDL Number MFCD00026667
InChI Key AFSIMBWBBOJPJG-UHFFFAOYSA-N
Synonym Stearic Acid Vinyl Ester
PubChem CID 66077
IUPAC Name ethenyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC=C

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 310.52
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Vinyl Stearate (stabilized with MEHQ)
EINECSNumber (2)-3474
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.