Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Vinyl Tape

Catalog No. 19070918 Shop All 3M Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-070-918 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-918 Supplier 3M™ Supplier No. 7000028852
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M Company Vinyl Tape is a rubber adhesive that provides excellent holding strength on many surfaces

  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • One-piece, clean removal from many surfaces which helps reduce clean-up and labor costs
  • Good abrasion and solvent resistance for long service life
  • Available in a wide variety of vivid colors for diverse applications
  • Floor and safety marking, color coding and industrial applications
  • Designed to resist wear, scrapes and moisture
  • 3M Catalog# 471
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W) 36 yd. x 2 in.
Packaging Quantity 24/Pk.
Thickness 5.2 mil
Length (Metric) 33 m
For Use With (Application) Paint masking; General use vinyl; Safety and hazard marking; Sealing plate edges against ink penetration
Width (Metric) 50 mm
Adhesive Type Rubber
Color Yellow
Material Vinyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.