Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Vinyl Tape

Catalog No. p-7182400 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
36 yd. x 1/2 in.
36 yd. x 1/4 in.
36 yd. x 2 in.
Quantity:
12 Rolls/Pk. 6 Pk./Cs.
144 Rolls/Cs.
24 Rolls/Cs.
72 Rolls/Cs.
Color:
Blue
Green
Orange
Red
White
Yellow
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Quantity Color
19-072-021 36 yd. x 1/4 in. 144 Rolls/Cs. Blue
19-072-042 36 yd. x 1/2 in. 12 Rolls/Pk. 6 Pk./Cs. Blue
19-072-103 36 yd. x 1/4 in. 144 Rolls/Cs. Green
19-072-070 36 yd. x 2 in. 24 Rolls/Cs. Green
19-072-078 36 yd. x 1/4 in. 144 Rolls/Cs. Orange
19-072-045 36 yd. x 1/2 in. 72 Rolls/Cs. Orange
19-072-058 36 yd. x 2 in. 24 Rolls/Cs. Orange
19-072-015 36 yd. x 1/4 in. 144 Rolls/Cs. Red
19-072-041 36 yd. x 1/2 in. 72 Rolls/Cs. Red
19-072-093 36 yd. x 2 in. 24 Rolls/Cs. Red
19-072-044 36 yd. x 1/2 in. 72 Rolls/Cs. White
19-072-016 36 yd. x 1/4 in. 144 Rolls/Cs. Yellow
19-072-043 36 yd. x 1/2 in. 72 Rolls/Cs. Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 19-072-021 Supplier 3M™ Supplier No. 7000048524
Only null left
Add to Cart
Add to Cart

3M™ Vinyl tape solves a variety of challenges, including floor and safety marking, color coding and industrial applications.

  • Available in a wide variety of vivid colors for diverse applications
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • Designed to resist wear, scrapes and moisture
  • One-piece, clean removal from many surfaces which helps reduce clean-up and labor costs
  • Rubber adhesive provides excellent holding strength on many surfaces
  • Good abrasion and solvent resistance for long service life
TRUSTED_SUSTAINABILITY

Specifications

Product Type Vinyl Tape
For Use With (Application) Color Coding Tools and Equipment, Electroplating and Anodizing Processes, Fine Line Masking, Floor
Adhesive Type Rubber
Color Blue
Dimensions (L x W) 36 yd. x 1/4 in.
Material Rubber
Model 471
Quantity 144 Rolls/Cs.
Thickness (English) 5.2 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.