Learn More
MSC VINYL TAPE
Catalog No. 5000794344
Encompass
This vinyl masking tape features a rubber adhesive, offering durable and flexible performance across a wide operating temperature range.
  • Made of vinyl material with a rubber adhesive
  • Measures 36 yd in length
  • Offers 5.2 mil thickness
  • Yellow color for easy identification
  • Tensile strength of 14 lb/in
  • Operating temperature range from 4°C (40°F) to 77°C (170°F)
  • General purpose protection, wrapping, and sealing
  • Suitable for die-cutting, bundling, and reinforcing
  • Effective for paint masking and identification
  • Applicable for anodizing and electroplating
  • Can be used as printable labels
  • Ideal for floor lane marking and color coding
Catalog No. 50-007-94344 Supplier MSC Supplier No. 43666940
This vinyl masking tape features a rubber adhesive, offering durable and flexible performance across a wide operating temperature range.
  • Made of vinyl material with a rubber adhesive
  • Measures 36 yd in length
  • Offers 5.2 mil thickness
  • Yellow color for easy identification
  • Tensile strength of 14 lb/in
  • Operating temperature range from 4°C (40°F) to 77°C (170°F)
  • General purpose protection, wrapping, and sealing
  • Suitable for die-cutting, bundling, and reinforcing
  • Effective for paint masking and identification
  • Applicable for anodizing and electroplating
  • Can be used as printable labels
  • Ideal for floor lane marking and color coding