Learn More
MSC VINYL TAPE
Catalog No. 5000685646
Encompass
3M Vinyl Tape 471 is a conformable, colored tape made from a vinyl backing with a rubber adhesive, suitable for a variety of industrial and scientific applications such as lane and safety markings, color coding, abrasion protection, masking, sealing, and splicing.
  • Pigmented backings maintain vivid colors even when exposed to heavy abrasion
  • Conformable backing with dead stretch properties for irregular surfaces
  • Rubber adhesive provides strong adhesion and holding strength
  • Sharp colors enhance plant safety and color coding systems
  • Clean removal from many surfaces reduces clean-up and labor costs
  • Abrasion and solvent resistant for extended application life
  • Low leachable halogens and sulfur for corrosion-sensitive applications
Catalog No. 50-006-85646 Supplier MSC Supplier No. 00048769
3M Vinyl Tape 471 is a conformable, colored tape made from a vinyl backing with a rubber adhesive, suitable for a variety of industrial and scientific applications such as lane and safety markings, color coding, abrasion protection, masking, sealing, and splicing.
  • Pigmented backings maintain vivid colors even when exposed to heavy abrasion
  • Conformable backing with dead stretch properties for irregular surfaces
  • Rubber adhesive provides strong adhesion and holding strength
  • Sharp colors enhance plant safety and color coding systems
  • Clean removal from many surfaces reduces clean-up and labor costs
  • Abrasion and solvent resistant for extended application life
  • Low leachable halogens and sulfur for corrosion-sensitive applications