Learn More
Scientific Commodities Inc VINYL TUBING .027X.045 100'
Catalog No. NC9014667
Encompass_Preferred
MICRO MEDICAL GRADE CATHETER VINYL TUBING,DIMENSIONS ID in. x OD in. -0.027 x 0.045,FOR SAFE USE WITH PLASMA, BLOOD, INTRAVENOUS, INTRACEREBRAL AND IMPLANTATION SOLUTIONS.
Catalog No. NC9014667 Supplier Scientific Commodities Inc Supplier No. BB31785V/3A
MICRO MEDICAL GRADE CATHETER VINYL TUBING,DIMENSIONS ID in. x OD in. -0.027 x 0.045,FOR SAFE USE WITH PLASMA, BLOOD, INTRAVENOUS, INTRACEREBRAL AND IMPLANTATION SOLUTIONS.