Learn More
AVANTOR MASTERFLEX (VWR LLC) Vinyl Tubing, 1/4""ID x 3/8""OD, 50 ft/pack
Catalog No. NC9001318
Encompass
Vinyl Tubing, 1/4"ID x 3/8"OD, 50 ft/pack, Applications: General-purpose lab use.Characteristics: Fair chemical and abrasion resistance, odorless, nontoxic, and transparent.Certification: Tubing is manufactured according to GMP.Temperature range: ¿45 to 180¿F(¿43 to 82¿C)Sterilize: Ethylene oxide.
Catalog No. NC9001318 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0640506
Vinyl Tubing, 1/4"ID x 3/8"OD, 50 ft/pack, Applications: General-purpose lab use.Characteristics: Fair chemical and abrasion resistance, odorless, nontoxic, and transparent.Certification: Tubing is manufactured according to GMP.Temperature range: ¿45 to 180¿F(¿43 to 82¿C)Sterilize: Ethylene oxide.