Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vinylboronic Acid Pinacol Ester

Catalog No. 6333481G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-334-81G 1 g
63-334-810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 63-334-81G Supplier Sigma Aldrich Supplier No. 6333481G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used forSuzuki-Miyaura coupling reactionsMizoroki-Heck reactions (cascade reaction)Intramolecular Nozaki-Hiyama-Kishi reactionsStereoselective Cu-catalyzed +a-selective and stereospecific couplingControl of stereoselectivity and mechanistic portrait on intramolecular (4+1)-cycloaddition of dialkoxycarbenesRegio- and stereoselective synthesis of trisubstituted alkenes via gold(I)-catalyzed hydrophosphoryloxylation of haloalkynes followed by Pd-catalyzed consecutive cross-coupling reactionsAsymmetric Birch reductive alkylationReagent used in Preparation of Molecular tubes for lipid sensingEnzymatic inhibitors, antibiotics, receptor analogs, and other biologically significant compounds (including total syntheses)

Specifications

CAS 75927-49-0
Density 0.908 g/mL at 25°C
Molecular Formula C8H15BO2
Refractive Index n20/D 1.4300
Linear Formula C8H15BO2
MDL Number MFCD00192492
Quantity 1 g
Synonym 2-Ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; 2-Vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaoborolane; 4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane
Molecular Weight (g/mol) 154.01 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.