Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vinylboronic Anhydride Pyridine Complex

Catalog No. 6379985G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-799-85G 5 g
63-799-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 63-799-85G Supplier Sigma Aldrich Supplier No. 6379985G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used forSuzuki-Miyaura cross-couplingStereoselective synthesis via Palladium-catalyzed carboamination Alkyl-connected 2-amino-6-vinylpurine (AVP) crosslinking agent to cytosine base in RNA Kaiser oxime resin-derived palladacycle as a recoverable polymeric precatalyst in Suzuki-Miyaura cross-coupling reactions in aqueous media Kinetic resolution of phosphoryl and sulfonyl esters of binaphthol derivatives via Pd-catalyzed alcoholysis of their vinyl ethers Stereoselective isomerization of N-allyl aziridines into Z-enamines by using rhodium hydride catalysisKinetic resolution of axially chiral biaryl derivatives via palladium/chiral diamine ligand-catalyzed alcoholysis Transition metal-catalyzed alkenylation of aziridines, cycloaddition and thermal rearrangement reactions Intramolecular Heck reaction strategy for synthesis of functionalized tetrahydroanthracenes Reagent used for Preparation ofBACE-1 inhibitors and SAR of cyclic sulfone hydroxyethylamines Distorted spiropentanes Small molecule bradykinin B2 receptor antagonists in angioedema therapy Enol Ethers Styryl cyclobutanone

Specifications

CAS 442850-89-7
Molecular Formula C6H9B3O3 -+ C5H5N
Linear Formula C6H9B3O3 -+ C5H5N
Quantity 5 g
MDL Number MFCD03839940
Synonym 2,4,6-Trivinylcyclotriboroxane pyridine complex; Trivinyl-boroxin pyridine complex; Trivinyl-boroxin pyridine complex; Trivinylcyclotriboroxane pyridine complex
Molecular Weight (g/mol) 240.67 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.