Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinylcyclopentane 97.0+%, TCI America™
SDP

Catalog No. V00791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V00791G Supplier TCI America Supplier No. V00791G

Chemical Identifiers

CAS 3742-34-5
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00001386
InChI Key BEFDCLMNVWHSGT-UHFFFAOYSA-N
Synonym vinylcyclopentane, cyclopentane, ethenyl, cyclopentane, vinyl, cyclopentyl-ethene, cyclopentylethylene, cyclopentane,ethenyl, acmc-209iss
PubChem CID 77345
IUPAC Name ethenylcyclopentane
SMILES C=CC1CCCC1

Specifications

Color Colorless
Boiling Point 100°C
Quantity 1 g
UN Number 3295
Formula Weight 96.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinylcyclopentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.