Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vinylene carbonate solution

Catalog No. 90159525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-159-525ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-159-525ML Supplier Sigma Aldrich Supplier No. 90159525ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

cis-(-)-Limonene oxide is a valuable chiral terpene oxide building block.Diastereoselective ring opening of limonene oxide with water catalysed by beta-ketophosphonate complexes of molybdenum(VI)

Specifications

CAS 872-36-6
Density 0.915 g/mL
Refractive Index n/D 1.377
Linear Formula C3H2O3
Quantity 5 g
Synonym 1,3-Dioxol-2-one; VC
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.