Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vinylmagnesium Bromide Solution

Catalog No. 257257200ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
200 mL
1.8 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-725-7200ML 200 mL
25-725-718L 1.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 25-725-7200ML Supplier Sigma Aldrich Supplier No. 257257200ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Storage below 25°C may cause formation of crystalline magnesium salts. Moving container to a war mLocation and occasional gentle swirling should redissolve the solid

Specifications

CAS 1826-67-1
Density 0.981 g/mL at 25°C
Molecular Formula C2H3BrMg
Linear Formula CH2=CHMgBr
MDL Number MFCD00000042
Quantity 200 mL
Molecular Weight (g/mol) 131.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.