Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vinylmagnesium Bromide Solution

Catalog No. 503081472 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
4 x 100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1472 4 x 100 mL
22-558-425ML 4 x 25 mL
22-558-4800ML 800 mL
22-558-4100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 50-308-1472 Supplier Sigma Aldrich Supplier No. 2255844X100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Vinylmagnesium bromide solution (1 M in THF) may be used in the synthesis of 2-vinylchroman-4-ones by the conjugate addition with chromones in the presence of a Lewis acid.It may also be used in the following transformations: (S)-3-(benzyloxy)-4-oxo-butanoic acid methyl ester to methyl (3S,4S)-4-hydroxy-3-(phenylmethoxy)hex-5-enoate.™-1-Azido-5,6-epoxyhexane to (S)-1-(4-azidobutyl)-2-propen-1-ol.2,3-O-Isopropylidene-D-ribose to 1-[(4R,5S)-5-((1S)-1-hydroxyallyl)-2,2-dimethyl[1,3]dioxolan-4-yl]ethane-1,2-diol.(4S,5S)-2,2-Dimethyl-5-vinyl[1,3]dioxolane-4-carbaldehyde to (1R,4R,5S)- and (1S,4R,5S)-1-(2,2-dimethyl-5-vinyl[1,3]dioxolan-4-yl)prop-2-en-1-ol.Ethyl levulinate to ?-methyl-?-vinylbutyrolactone.Storage below 25°C may cause crystallization. War mgently to redissolve.

Specifications

CAS 1826-67-1
Density 0.981 g/mL at 25°C
Molecular Formula C2H3BrMg
Linear Formula CH2=CHMgBr
Quantity 4 x 100 mL
MDL Number MFCD00000042
Molecular Weight (g/mol) 131.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.