Learn More
Strem, An Ascensus Company CAS# 1746-03-8. 250g. Vinylphosphonic acid, min. 90%. MFCD00043866
SDP
Catalog No. 503712
Encompass
CAS# 1746-03-8. 250g. Vinylphosphonic acid, min. 90%. MFCD00043866. Molecular Weight: 108.00. Molecular Formula: CH2=CHP(O)(OH)2. Color/form: colorless to pale-yellow liq. Strem# 15-9155. http://www.strem.com/catalog/v/15-9155/
Catalog No. 50-371-2 Supplier Strem, An Ascensus Company Supplier No. 159155250G
CAS# 1746-03-8. 250g. Vinylphosphonic acid, min. 90%. MFCD00043866. Molecular Weight: 108.00. Molecular Formula: CH2=CHP(O)(OH)2. Color/form: colorless to pale-yellow liq. Strem# 15-9155. http://www.strem.com/catalog/v/15-9155/