Learn More
Cytek Biosciences Inc vF 450 aMu CD24 Anti Primary
Supplier: Cytek Biosciences Inc 750242U100
Product Name: violetFluor 450 Anti-Mouse CD24 (M1/69)
Product Class: Antibody
Type: Primary
Product Name: violetFluor 450 Anti-Mouse CD24 (M1/69)
Cytek Catalog Number: 75-0242-U100
Marker: CD24
Target Species: Mouse
Clonality: Monoclonal
Clone: M1/69
Host Species: Rat
Isotype: IgG2b, k
Conjugation: violetFluor 450
Size: 100 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.