Learn More
AdipoGen Violacein,Chemical. CAS: 548-54-9
SDP
Catalog No. 501176239
Encompass_Preferred
Violacein, AdipoGen Life Sciences. Chemical. CAS: 548-54-9. Formula: C20H13N3O3. MW: 343.3. Isolated from Chromobacterium violaceum. Antibacterial, antifungal and antiprotozoal agent. Quorum sensing metabolite. Antitumor compound. Protective against UV-C irradiation. Possesses immunomodulatory, analgesic and antipyretic activity. Indole-derived purple-colored pigment (free of deoxyviolacein). 548-54-9
Catalog No. 50-117-6239 Supplier AdipoGen Supplier No. BVT0473M001
Violacein, AdipoGen Life Sciences. Chemical. CAS: 548-54-9. Formula: C20H13N3O3. MW: 343.3. Isolated from Chromobacterium violaceum. Antibacterial, antifungal and antiprotozoal agent. Quorum sensing metabolite. Antitumor compound. Protective against UV-C irradiation. Possesses immunomodulatory, analgesic and antipyretic activity. Indole-derived purple-colored pigment (free of deoxyviolacein). 548-54-9